missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dimethyl Sulfoxide, For headspace gas chromatography SupraSolv™ MilliporeSigma™
For headspace gas chromatography SupraSolv™
$341.24 - $1640.66
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
IUPAC Name | methanesulfinylmethane |
SMILES | CS(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
M1019000500
|
Technidata
1019000500 |
500 mL | Glass Bottle |
Each for $341.24
|
|
|||||
M1019001000
|
Technidata
1019001000 |
1 L | Glass Bottle |
Each for $790.97
|
|
|||||
M1019002500
|
Technidata
1019002500 |
2.5 L | Glass Bottle |
Each for $1,640.66
|
|
|||||
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methanesulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 | |
CS(C)=O |
Specifications
18.5°C | |
Colorless | |
189°C | |
87°C (189°F) | |
Charactristic | |
Liquid | |
C2H6OS | |
0.6 hPa (20°C) | |
Soluble in water: 1000g/L at 20°C | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
Dimethyl Sulfoxide |
67-68-5 | |
1.10g/cm3 at 20°C | |
500 mL | |
300°to302°C (572°to576°F) | |
Glass Bottle | |
1.48 | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methanesulfinylmethane | |
679 | |
78.13 |
Safety and Handling
Recommended Storage : 15° to 25°C (59° to 77°F); Tightly closed. Dry