Learn More
Dimethyl trisulfide, 98%, Thermo Scientific Chemicals
$213.51 - $635.51
Chemical Identifiers
CAS | 3658-80-8 |
---|---|
Molecular Formula | C2H6S3 |
Molecular Weight (g/mol) | 126.25 |
MDL Number | MFCD00039808 |
InChI Key | YWHLKYXPLRWGSE-UHFFFAOYSA-N |
Synonym | dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts |
PubChem CID | 19310 |
ChEBI | CHEBI:4614 |
IUPAC Name | (methyltrisulfanyl)methane |
SMILES | CSSSC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH3291306
|
Thermo Scientific Chemicals
H3291306 |
5 g |
Each for $213.51
|
|
|||||
AAH3291314
|
Thermo Scientific Chemicals
H3291314 |
25 g |
Each for $635.51
|
|
|||||
Description
Dimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethyl trisulfide is reported as a major aroma component in cooked Brassicaceous vegetables. Trap baits containing dimethyl trisulfide have been used to capture Calliphora loewi and other blowflies. Dimethyl trisulfide along with dimethyl sulfide and dimethyl disulfide have been confirmed as volatile compounds given off by the fly-attracting plant known as dead-horse arum (Helicodiceros muscivorus). These flies are attracted to the odor of fetid meat and help pollinate this plant.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C. Store in an air-tight and light-resistant container.
Chemical Identifiers
3658-80-8 | |
126.25 | |
YWHLKYXPLRWGSE-UHFFFAOYSA-N | |
19310 | |
(methyltrisulfanyl)methane |
C2H6S3 | |
MFCD00039808 | |
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts | |
CHEBI:4614 | |
CSSSC |
Specifications
3658-80-8 | |
165°C to 175°C | |
Stench | |
1.6 | |
MFCD00039808 | |
UN1993 | |
Insoluble in water. | |
CSSSC | |
126.25 | |
CHEBI:4614 | |
98% |
1.203 | |
52°C (126°F) | |
C2H6S3 | |
CH3SSSCH3 | |
5 g | |
dimethyl trisulfide, trisulfide, dimethyl, methyl trisulfide, 2,3,4-trithiapentane, dimethyl trisulphide, dimethyltrisulfide, methyltrisulfanyl methane, dimethyltrisulfane, unii-3e691t3nl1, dmts | |
YWHLKYXPLRWGSE-UHFFFAOYSA-N | |
(methyltrisulfanyl)methane | |
19310 | |
126.26 | |
Dimethyl trisulfide |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 222-910-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only