missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117100025

 View more versions of this product

Catalog No. AC117100025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dioctyl sulfosuccinate, sodium salt
2.5 max. (on solids basis)
White
95% min
C20H37NaO7S
2.5 kg
15,149
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
23673837
96%
Waxy Solid
577-11-7
153.0°C to 157.0°C
Authentic
Plastic drum
MFCD00012455
04, IV, 114
15, 3446
Solubility in water: 1.5g/100mL (25°C). Other solubilities: soluble in ccl4,petroleum ether,xylene and,alcohol
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
444.56
444.55
300ppm (50% max. in toluene)

Chemical Identifiers

577-11-7
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
23673837
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
C20H37NaO7S
MFCD00012455
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Danger

EINECSNumber : 209-406-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only