missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals

$84.17 - $502.95

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
PubChem CID 23673837
IUPAC Name sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC117101000
SDS View Documents
Thermo Scientific Chemicals
117101000
100 g Plastic Bottle
Each for $84.17
 
AC117100010
SDS View Documents
Thermo Scientific Chemicals
117100010
1 kg Plastic Bottle
Each for $312.85
 
AC117100025
SDS View Documents
Thermo Scientific Chemicals
117100025
2.5 kg Plastic drum
Each for $502.95
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

577-11-7
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
23673837
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
C20H37NaO7S
MFCD00012455
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate

Specifications

2.5 max. (on solids basis)
White
96%
C20H37NaO7S
100 g
15,149
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
23673837
≥95%
Dioctyl sulfosuccinate, sodium salt
577-11-7
Authentic
Plastic Bottle
MFCD00012455
04, IV, 114
15, 3446
300ppm max. Insoluble Matter (in toluene, in 50% soln.)
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
444.56
444.55
Waxy Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Danger

EINECSNumber : 209-406-4

SDS
Documents

Documents

RUO – Research Use Only