missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dioctyl sulfosuccinate, sodium salt, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 117101000

 View more versions of this product

Catalog No. AC117101000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dioctyl sulfosuccinate, sodium salt
577-11-7
Authentic
Plastic Bottle
MFCD00012455
04, IV, 114
15, 3446
300ppm max. Insoluble Matter (in toluene, in 50% soln.)
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
444.56
444.55
Waxy Solid
2.5 max. (on solids basis)
White
96%
C20H37NaO7S
100 g
15,149
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
23673837
≥95%

Chemical Identifiers

577-11-7
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
23673837
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
C20H37NaO7S
MFCD00012455
docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only