missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenhydramine hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1013618

 View more versions of this product

Catalog No. AAA1013618


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A histamine H1 receptor antagonistDiphenhydramine hydrochloride acts as a sodium channel blocker. It is used as a histamine H1-receptor antagonist. Further, it is used as a sedative, hypnotic, antiemetic, antitussive and pruritus. It is employed in the treatment of allergic conditions. In addition to this, it is used as an active ingredient in common cold preparations.

Solubility
Soluble in water, dimethyl sulfoxide, dimethylformamide, methanol and ethanol.

Notes
Incompatible with strong oxidizing agents, strong acids and alkalis.
Specifications

Specifications

Diphenhydramine hydrochloride
168°C to 169°C
MFCD00012479
UN2811
diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
PCHPORCSPXIHLZ-UHFFFAOYSA-N
hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
8980
291.82
147-24-0
C17H22ClNO
50 g
Light sensitive
Soluble in water,dimethyl sulfoxide,dimethylformamide,methanol and ethanol.
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
291.82
CHEBI:4637
99%

Chemical Identifiers

147-24-0
291.82
PCHPORCSPXIHLZ-UHFFFAOYSA-N
8980
hydrogen [2-(diphenylmethoxy)ethyl]dimethylamine chloride
C17H22ClNO
MFCD00012479
diphenhydramine hydrochloride, dimedrol, diphenhydramine hcl, carphenamine, bendylate, carphenex, cathejell, denydryl, diphamine, eldadryl
CHEBI:4637
[H+].[Cl-].CN(C)CCOC(C1=CC=CC=C1)C1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H361-H302-H315-H319-H335
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-687-2

RTECSNumber : KR7000000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only