missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenyl phosphate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 225690050

 View more versions of this product

Catalog No. AC225690050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diphenyl phosphate
838-85-7
White
99%
C12H11O4P
MFCD00003033
06, 178
ASMQGLCHMVWBQR-UHFFFAOYSA-N
diphenyl hydrogen phosphate
13282
99%
99%
63°C to 70°C
Authentic
Glass bottle
(C6H5O)2P(O)OH
5 g
diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
250.19
250.19
Crystalline Powder

Chemical Identifiers

838-85-7
250.19
ASMQGLCHMVWBQR-UHFFFAOYSA-N
13282
OP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
C12H11O4P
MFCD00003033
diphenyl phosphate, phosphoric acid, diphenyl ester, phenyl hydrogen phosphate, phenyl phosphate pho 2 ho po, diphenoxyphosphinic acid, phosphoric acid diphenyl ester, diphenylphosphoric acid, phosphoric acid diphenyl, dsstox_cid_28182
diphenyl hydrogen phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 212-657-2

RTECSNumber : TC5470000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only