missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenylacetylene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 117180050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diphenylacetylene | |
58.0°C to 61.0°C | |
0.9900g/mL | |
Authentic | |
Glass bottle | |
C6H5C≡CC6H5 | |
05,656 | |
0.99 | |
diphenylacetylene, tolan, diphenylethyne, 1,2-diphenylethyne, tolane, 1,2-diphenylacetylene, benzene, 1,1'-1,2-ethynediyl bis, biphenylacetylene, diphenyl acetylene, ethyne, diphenyl | |
JRXXLCKWQFKACW-UHFFFAOYSA-N | |
2-phenylethynylbenzene | |
10390 | |
178.23 | |
Crystalline Powder and/or Chunks |
501-65-5 | |
Brown to Yellow | |
170.0°C (19.0 mmHg) | |
98.5% min. (GC) | |
C14H10 | |
5 g | |
01,335 | |
15,9664 | |
Solubility in water: insoluble. Other solubilities: soluble in ether and hot alcohol | |
C1=CC=C(C=C1)C#CC2=CC=CC=C2 | |
178.23 | |
CHEBI:51579 | |
99% |
Chemical Identifiers
501-65-5 | |
178.23 | |
diphenylacetylene, tolan, diphenylethyne, 1,2-diphenylethyne, tolane, 1,2-diphenylacetylene, benzene, 1,1'-1,2-ethynediyl bis, biphenylacetylene, diphenyl acetylene, ethyne, diphenyl | |
CHEBI:51579 | |
C1=CC=C(C=C1)C#CC2=CC=CC=C2 |
C14H10 | |
JRXXLCKWQFKACW-UHFFFAOYSA-N | |
10390 | |
2-phenylethynylbenzene |
Safety and Handling
EINECSNumber : 207-926-6
RUO – Research Use Only