missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diphenylcarbazone-Bromophenol Blue, For Chloride, Certified, LabChem™

Supplier:  LabChem LC136801

 View more versions of this product

Catalog No. LC136801

Explore available promotions

Description

Description

Specifications

Specifications

Diphenylcarbazone-Bromophenol Blue
99.45,0.5,0.05
C13H14N4O
Soluble in water
242.28
Amber Poly
Sulfur compounds; Carbon monoxide; Carbon dioxide; May release flammable gases; Nitrogen oxides; Hydrogen bromide
500 mL
For Chloride
538-62-5,538-62-5,140-22-7,115-39-9
UN1993
Passes Test
Certified
Passes Test
Orange
Liquid
SDS
Documents

Documents

Promotions

Promotions