missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diphenylcarbazone-Bromophenol Blue, For Chloride, Certified, LabChem™
$82.53 - $287.91
Specifications
Chemical Name or Material | Diphenylcarbazone-Bromophenol Blue |
---|---|
Name Note | For Chloride |
CAS Max % | 99.45,0.5,0.05 |
CAS | 538-62-5,538-62-5,140-22-7,115-39-9 |
Linear Formula | C13H14N4O |
Specifications
Diphenylcarbazone-Bromophenol Blue | |
99.45,0.5,0.05 | |
C13H14N4O | |
Soluble in water | |
242.28 | |
Passes Test | |
Orange |
For Chloride | |
538-62-5,538-62-5,140-22-7,115-39-9 | |
UN1993 | |
Passes Test | |
Certified | |
Sulfur compounds; Carbon monoxide; Carbon dioxide; May release flammable gases; Nitrogen oxides; Hydrogen bromide | |
Liquid |