missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dipropyl Phthalate 98.0+%, TCI America™
$126.40 - $981.57
Chemical Identifiers
CAS | 131-16-8 |
---|---|
Molecular Formula | C14H18O4 |
Molecular Weight (g/mol) | 250.294 |
MDL Number | MFCD00009371 |
InChI Key | MQHNKCZKNAJROC-UHFFFAOYSA-N |
Synonym | dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss |
PubChem CID | 8559 |
ChEBI | CHEBI:60069 |
IUPAC Name | dipropyl benzene-1,2-dicarboxylate |
SMILES | CCCOC(=O)C1=CC=CC=C1C(=O)OCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
P030625ML
|
TCI America
P030625ML |
25 mL |
Each for $126.40
|
|
||||||
P0306100ML
|
TCI America
P0306100ML |
100 mL |
Each for $375.25
|
|
||||||
P0306500ML
|
TCI America
P0306500ML |
500 mL |
Each for $981.57
|
|
||||||
Chemical Identifiers
131-16-8 | |
250.294 | |
MQHNKCZKNAJROC-UHFFFAOYSA-N | |
8559 | |
dipropyl benzene-1,2-dicarboxylate |
C14H18O4 | |
MFCD00009371 | |
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss | |
CHEBI:60069 | |
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC |
Specifications
131-16-8 | |
305°C | |
MFCD00009371 | |
3082 | |
MQHNKCZKNAJROC-UHFFFAOYSA-N | |
dipropyl benzene-1,2-dicarboxylate | |
8559 | |
250.29 | |
Liquid |
Yellow | |
C14H18O4 | |
25 mL | |
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss | |
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC | |
250.294 | |
CHEBI:60069 | |
≥98.0% (GC) | |
Dipropyl Phthalate |
Safety and Handling
RTECSNumber : TI1940000
TSCA : Yes