missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dipropyl Phthalate 98.0+%, TCI America™
Supplier: TCI America P0306500ML
Specifications
Dipropyl Phthalate | |
Yellow | |
C14H18O4 | |
500 mL | |
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss | |
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC | |
250.294 | |
CHEBI:60069 | |
≥98.0% (GC) |
131-16-8 | |
305°C | |
MFCD00009371 | |
3082 | |
MQHNKCZKNAJROC-UHFFFAOYSA-N | |
dipropyl benzene-1,2-dicarboxylate | |
8559 | |
250.29 | |
Liquid |
Chemical Identifiers
131-16-8 | |
250.294 | |
MQHNKCZKNAJROC-UHFFFAOYSA-N | |
8559 | |
dipropyl benzene-1,2-dicarboxylate |
C14H18O4 | |
MFCD00009371 | |
dipropyl phthalate, di-n-propyl phthalate, di-n-propylphthalate, phthalic acid dipropyl ester, phthalic acid, dipropyl ester, phthalic acid di-n-propyl ester, 1,2-benzenedicarboxylic acid, dipropyl ester, dipropylphthalate, n-dipropyl phthalate, unii-8usp5y77ss | |
CHEBI:60069 | |
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC |
Safety and Handling
RTECSNumber : TI1940000
TSCA : Yes