Learn More
Disodium terephthalate, 99+%, Thermo Scientific Chemicals
$49.62 - $1031.03
Chemical Identifiers
CAS | 10028-70-3 |
---|---|
Molecular Formula | C8H4Na2O4 |
Molecular Weight (g/mol) | 210.096 |
MDL Number | MFCD00013137 |
InChI Key | VIQSRHWJEKERKR-UHFFFAOYSA-L |
Synonym | disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:? |
PubChem CID | 82305 |
IUPAC Name | disodium;terephthalate |
SMILES | C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4294606
|
Thermo Scientific Chemicals
04294606 |
5 g |
Each for $49.62
|
|
|||||
AA4294614
|
Thermo Scientific Chemicals
04294614 |
25 g |
Each for $97.58
|
|
|||||
AA4294622
|
Thermo Scientific Chemicals
04294622 |
100 g |
Each for $229.49
|
|
|||||
AA4294636
|
Thermo Scientific Chemicals
04294636 |
500 g |
Each for $1,031.03
|
|
|||||
Description
Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10028-70-3 | |
210.096 | |
VIQSRHWJEKERKR-UHFFFAOYSA-L | |
82305 | |
C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+] |
C8H4Na2O4 | |
MFCD00013137 | |
disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:? | |
disodium;terephthalate |
Specifications
10028-70-3 | |
MFCD00013137 | |
disodium terephthalate, terephthalic acid disodium salt, terephthalic acid, disodium salt, sodium terephthalate, unii-gi30zoc0oo, gi30zoc0oo, disodiumterephthalate, acmc-1bvw0, disodium terephthalate 5g, 1,4-benzenedicarboxylicacid, sodium salt 1:? | |
VIQSRHWJEKERKR-UHFFFAOYSA-L | |
disodium;terephthalate | |
82305 | |
≥99% | |
Disodium terephthalate |
C8H4Na2O4 | |
5 g | |
Soluble in water. | |
C1=CC(=CC=C1C(=O)[O-])C(=O)[O-].[Na+].[Na+] | |
210.096 | |
210.1 | |
Powder |
Safety and Handling
EINECSNumber : 233-075-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only