Disposable Aluminum Dishes
Withstand temperatures up to 677°C (1250.6°F).
Supplier:  Customer Channels Group (Ccg) 43100
 View more versions of this product
Catalog No. S40283
Explore available promotions
Description
Description
  • Designed for weighing, dispensing and storing
  • Can be used as evaporating dishes or as dust covers
  • With crimped sides for rigidity and a smooth bottom
Order Info
Sold in packs of 100.
Specifications
Specifications
Flat
Single-use
Round
Designed for weighing, dispensing and storing, can be used as evaporating dishes or as dust covers
4.39 cm (Inner Top)
20 mL
Aluminum
Weighing Dish
4.39 cm
SDS
Documents
Documents
Accessories