Cole-Parmer™ Essentials Disposable Inoculation Loops
Save time and eliminate cross-contamination
Supplier:  Cole-Parmer™ Essentials 0623104
 View more versions of this product
Catalog No. 03-391-561
Explore available promotions
Description
Description
  • Color coded
  • Sterile
Smooth and flexible loops are constructed of high-impact polystyrene. Loops ensure uniform streaking without damaging your gel surface. Color coding identifies inoculation volume easily. Packed sterile in tamperproof resealable bags. Each case is supplied with a quality certificate confirming sterility and accuracy.
Specifications
Specifications
Single-use
Polystyrene
10 μL
Inoculating Loop
Sterile
22.7 cm
Yellow
40