missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™

Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %

Supplier:  Fisher BioReagents BP17225

 View more versions of this product

Catalog No. BP172-25



Explore available promotions

Description

Description

Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %

  • Commonly used as reducing agent to disrupt disulfide bonds in proteins in SDS-PAGE
  • Functionally tested for use in electrophoresis
  • Tested for absence of DNase, RNase, and protease
  • DTT is also used as an enzyme stabilizer during protein purification and enzymatic reactions
Specifications

Specifications

Dithiothreitol
0.04 AU max.
White
125°C
≥99 %
C4H10O2S2
25 g
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
C(C(C(CS)O)O)S
154.24
CHEBI:42170
≥99%
Powder or Solid
3483-12-3
38°C
5.1
DNase-, RNase- and Protease-Free
Glass Bottle
HSCH2CH(OH)CH(OH)CH2SH
15, 3419
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.24
Electrophoresis

Chemical Identifiers

3483-12-3
154.24
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
CHEBI:42170
C(C(C(CS)O)O)S
C4H10O2S2
VHJLVAABSRFDPM-IMJSIDKUSA-N
446094
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
Videos
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS Documents

Documents

Promotions

Promotions