missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dithiothreitol (White Crystals or Powder/Electrophoresis), Fisher BioReagents™

Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %

$103.78 - $374.98

Chemical Identifiers

CAS 3483-12-3
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
InChI Key VHJLVAABSRFDPM-IMJSIDKUSA-N
Synonym dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
PubChem CID 446094
ChEBI CHEBI:42170
IUPAC Name (2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
SMILES C(C(C(CS)O)O)S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP172-5
SDS View Documents
Fisher BioReagents
BP1725
5 g Glass Bottle
Each for $103.78
Case of 6 Each for $622.68
 
BP172-25
SDS View Documents
Fisher BioReagents
BP17225
25 g Glass Bottle
Each for $374.98
Case of 6 Each for $2,249.88
 
Description

Description

Dithiothreitol, >-99%, C4H10O2S2, CAS Number-3483-12-3, 1,4-dithio-dl-threitol, 1,4-dithiothreitol, 2s,3s-1,4-dimercaptobutane-2,3-diol, d-1,4-dithiothreitol, d-dtt, dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, dtt, threo-1,4-dimercapto-2,3-butanediol, 5g, 0.04 AU max., >-99 %

Dithiothreitol is used to reduce disulfide bonds in proteins and peptides and is added in low concentrations as an enzyme stabilizer during protein purification and enzymatic reactions.
Specifications

Chemical Identifiers

3483-12-3
154.24
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
CHEBI:42170
C(C(C(CS)O)O)S
C4H10O2S2
VHJLVAABSRFDPM-IMJSIDKUSA-N
446094
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol

Specifications

3483-12-3
38°C
5.1
≥99 %
C4H10O2S2
5 g
dithiothreitol, dl-1,4-dithiothreitol, dl-dithiothreitol, 1,4-dithio-dl-threitol, d-1,4-dithiothreitol, d-dtt, 2s,3s-1,4-dimercaptobutane-2,3-diol, threo-1,4-dimercapto-2,3-butanediol, 1,4-dithiothreitol, dtt
C(C(C(CS)O)O)S
154.24
CHEBI:42170
≥99%
Powder or Solid
0.04 AU max.
White
125°C
Glass Bottle
HSCH2CH(OH)CH(OH)CH2SH
15, 3419
VHJLVAABSRFDPM-IMJSIDKUSA-N
(2S,3S)-1,4-bis(sulfanyl)butane-2,3-diol
446094
154.24
DNase-, RNase- and Protease-Free
Dithiothreitol
Safety and Handling

Safety and Handling

Recommended Storage : -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'