missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Divinyl sulfone, 97%, stab. with 0.05% hydroquinone, Thermo Scientific Chemicals

$124.73 - $387.15

Chemical Identifiers

CAS 77-77-0
Molecular Formula C4H6O2S
Molecular Weight (g/mol) 118.15
MDL Number MFCD00008623
InChI Key AFOSIXZFDONLBT-UHFFFAOYSA-N
Synonym divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
PubChem CID 6496
ChEBI CHEBI:53729
IUPAC Name 1-ethenylsulfonylethene
SMILES C=CS(=O)(=O)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1282709
View Documents
Thermo Scientific Chemicals
L1282709
10 g
Each for $124.73
 
AAL1282718
View Documents
Thermo Scientific Chemicals
L1282718
50 g
Each for $387.15
 
Description

Description

Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.
Specifications

Chemical Identifiers

77-77-0
118.15
AFOSIXZFDONLBT-UHFFFAOYSA-N
6496
1-ethenylsulfonylethene
C4H6O2S
MFCD00008623
divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
CHEBI:53729
C=CS(=O)(=O)C=C

Specifications

77-77-0
1.177
102°C (215°F)
1.476
10 g
1071329
Miscible with water.
C=CS(=O)(=O)C=C
118.15
CHEBI:53729
97%
∼-26°C
233°C to 234°C
C4H6O2S
MFCD00008623
UN2927
divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
AFOSIXZFDONLBT-UHFFFAOYSA-N
1-ethenylsulfonylethene
6496
118.16
Divinyl sulfone, stabilized with 0.05% hydroquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H310-H314-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 201-057-6

RTECSNumber : KM7175000

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only