missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dizocilpine maleate, 99+%, Thermo Scientific Chemicals

NMDA receptor antagonist that prevents calcium flux

Supplier:  Thermo Scientific Chemicals J63917MF

 View more versions of this product

Catalog No. AAJ63917MF


Explore available promotions

Description

Description

Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.

Solubility
Soluble in ethanol.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

Dizocilpine maleate
White
MFCD00082465
14,3386
Soluble in ethanol.
CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O
6420042
≥99%
77086-22-7
C20H19NO4
50 mg
dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126
QLTXKCWMEZIHBJ-PJGJYSAQSA-N
337.375
337.37
Crystalline Powder

Chemical Identifiers

77086-22-7
337.375
QLTXKCWMEZIHBJ-PJGJYSAQSA-N
6420042
C20H19NO4
MFCD00082465
dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126
CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 278-614-5

RTECSNumber : HP1093575

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only