Learn More
Dizocilpine maleate, 99+%, Thermo Scientific Chemicals
NMDA receptor antagonist that prevents calcium flux
$219.14 - $673.86
Chemical Identifiers
CAS | 77086-22-7 |
---|---|
Molecular Formula | C20H19NO4 |
Molecular Weight (g/mol) | 337.375 |
MDL Number | MFCD00082465 |
InChI Key | QLTXKCWMEZIHBJ-PJGJYSAQSA-N |
Synonym | dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126 |
PubChem CID | 6420042 |
SMILES | CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ63917MA
|
Thermo Scientific Chemicals
J63917MA |
10 mg |
Each for $219.14
|
|
|||||
AAJ63917MF
|
Thermo Scientific Chemicals
J63917MF |
50 mg |
Each for $673.86
|
|
|||||
Description
Dizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDizocilpine maleate is used to interact at the NMDA-associated ion channel, preventing Ca2+ flux and blocking NMDA-induced depolarizations. (+)-MK 801 demonstrates potent neuroprotective effects against excitotoxic insult, showing substantial protection against neuron loss produced by NMDA and quinolinic acid introduction.
Solubility
Soluble in ethanol.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
77086-22-7 | |
337.375 | |
QLTXKCWMEZIHBJ-PJGJYSAQSA-N | |
6420042 |
C20H19NO4 | |
MFCD00082465 | |
dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126 | |
CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O |
Specifications
77086-22-7 | |
C20H19NO4 | |
10 mg | |
dizocilpine maleate, +-mk-801 hydrogen maleate, +-mk 801 maleate, dizocilpine hydrogen maleate, unii-6lr8c1b66q, dizocilpine maleate usan, dsstox_cid_25785, dsstox_rid_81126 | |
QLTXKCWMEZIHBJ-PJGJYSAQSA-N | |
337.375 | |
337.37 | |
Crystalline Powder |
White | |
MFCD00082465 | |
14,3386 | |
Soluble in ethanol. | |
CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O | |
6420042 | |
≥99% | |
Dizocilpine maleate |
Safety and Handling
EINECSNumber : 278-614-5
RTECSNumber : HP1093575
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only