missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Phenylpropionaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130860250

 View more versions of this product

Catalog No. AC130860250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

DL-2-Phenylpropionaldehyde
-60.0°C
1.0110g/mL
76°C
97.5% min. (GC)
C9H10O
CH3CH(C6H5)CHO
25 g
1.011
Solubility in water: 3.4g/L (20°C).
CC(C=O)C1=CC=CC=C1
134.18
2.16 mPa.s (20°C)
98%
93-53-8
Colorless
202.0°C to 205.0°C
Authentic
Glass bottle
1.5160 to 1.5240
MFCD00006973
07, 305
2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
IQVAERDLDAZARL-UHFFFAOYSA-N
2-phenylpropanal
7146
134.18
Liquid

Chemical Identifiers

93-53-8
134.18
IQVAERDLDAZARL-UHFFFAOYSA-N
7146
CC(C=O)C1=CC=CC=C1
C9H10O
MFCD00006973
2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
2-phenylpropanal
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of damaging fertility.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti

GHS Signal Word: Warning

EINECSNumber : 202-255-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only