Learn More
DL-2-Phenylpropionaldehyde, 98%, Thermo Scientific Chemicals
$37.71 - $108.41
Chemical Identifiers
CAS | 93-53-8 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.18 |
MDL Number | MFCD00006973 |
InChI Key | IQVAERDLDAZARL-UHFFFAOYSA-N |
Synonym | 2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy |
PubChem CID | 7146 |
IUPAC Name | 2-phenylpropanal |
SMILES | CC(C=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC130860250
|
Thermo Scientific Chemicals
130860250 |
25 g | Glass bottle |
Each for $37.71
|
|
||||
AC130861000
|
Thermo Scientific Chemicals
130861000 |
100 g | Glass bottle |
Each for $108.41
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-53-8 | |
134.18 | |
IQVAERDLDAZARL-UHFFFAOYSA-N | |
7146 | |
CC(C=O)C1=CC=CC=C1 |
C9H10O | |
MFCD00006973 | |
2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy | |
2-phenylpropanal |
Specifications
93-53-8 | |
Colorless | |
202.0°C to 205.0°C | |
Authentic | |
Glass bottle | |
1.5160 to 1.5240 | |
MFCD00006973 | |
07, 305 | |
2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy | |
IQVAERDLDAZARL-UHFFFAOYSA-N | |
2-phenylpropanal | |
7146 | |
134.18 | |
Liquid |
-60.0°C | |
1.0110g/mL | |
76°C | |
97.5% min. (GC) | |
C9H10O | |
CH3CH(C6H5)CHO | |
25 g | |
1.011 | |
Solubility in water: 3.4g/L (20°C). | |
CC(C=O)C1=CC=CC=C1 | |
134.18 | |
2.16 mPa.s (20°C) | |
98% | |
DL-2-Phenylpropionaldehyde |
Safety and Handling
GHS H Statement
Suspected of damaging fertility.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protecti
GHS Signal Word: Warning
EINECSNumber : 202-255-5
RUO – Research Use Only