missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 232935000

 View more versions of this product

Catalog No. AC232935000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

DL-2-Pyrrolidone-5-carboxylic acid
0.3% max.
Authentic
Plastic bottle
MFCD00064322
dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
ODHCTXKNWHHXJC-UHFFFAOYNA-N
5-oxopyrrolidine-2-carboxylic acid
499
129.12
149-87-1
0.5% max.
98%
C5H7NO3
500g
Solubility in water: 57g/l (20°C). Other solubilities: soluble in alcohol and acetone
OC(=O)C1CCC(=O)N1
129.12
CHEBI:16010
98%

Chemical Identifiers

149-87-1
129.12
ODHCTXKNWHHXJC-UHFFFAOYNA-N
499
5-oxopyrrolidine-2-carboxylic acid
C5H7NO3
MFCD00064322
dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
CHEBI:16010
OC(=O)C1CCC(=O)N1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.

GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici

GHS Signal Word: Danger

EINECSNumber : 205-748-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only