missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 149-87-1 |
---|---|
Molecular Formula | C5H7NO3 |
Molecular Weight (g/mol) | 129.12 |
MDL Number | MFCD00064322 |
InChI Key | ODHCTXKNWHHXJC-UHFFFAOYNA-N |
Synonym | dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid |
PubChem CID | 499 |
ChEBI | CHEBI:16010 |
IUPAC Name | 5-oxopyrrolidine-2-carboxylic acid |
SMILES | OC(=O)C1CCC(=O)N1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC232935000
|
Thermo Scientific Chemicals
232935000 |
500g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
149-87-1 | |
129.12 | |
ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
499 | |
5-oxopyrrolidine-2-carboxylic acid |
C5H7NO3 | |
MFCD00064322 | |
dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid | |
CHEBI:16010 | |
OC(=O)C1CCC(=O)N1 |
Specifications
149-87-1 | |
0.5% max. | |
98% | |
C5H7NO3 | |
500g | |
Solubility in water: 57g/l (20°C). Other solubilities: soluble in alcohol and acetone | |
OC(=O)C1CCC(=O)N1 | |
129.12 | |
CHEBI:16010 | |
98% |
0.3% max. | |
Authentic | |
Plastic bottle | |
MFCD00064322 | |
dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid | |
ODHCTXKNWHHXJC-UHFFFAOYNA-N | |
5-oxopyrrolidine-2-carboxylic acid | |
499 | |
129.12 | |
DL-2-Pyrrolidone-5-carboxylic acid |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 205-748-3
RUO â Research Use Only