missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-alpha-Tocopherol 96.0+%, TCI America™

$57.10 - $212.81

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T025125G
View Documents
TCI America
T025125G
25 g
Each for $57.10
 
T0251250G
View Documents
TCI America
T0251250G
250 g
Each for $212.81
 
Description

Description

Specifications

Chemical Identifiers

10191-41-0
430.72
GVJHHUAWPYXKBD-IEOSBIPESA-N
14985
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
C29H50O2
MFCD00072051
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
CHEBI:18145
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

10191-41-0
Brown-Yellow
C29H50O2
25 g
GVJHHUAWPYXKBD-IEOSBIPESA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydro-2H-1-benzopyran-6-ol
14985
430.72
Liquid
4°C
220°C
MFCD00072051
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
430.72
CHEBI:18145
≥96.0% (GC)
DL-alpha-Tocopherol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0864

RTECSNumber : GA8746000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'