missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Isoproterenol hydrochloride, 98%, Thermo Scientific Chemicals

Standard selective beta-adrenoceptor agonist

Supplier:  Thermo Scientific Chemicals J6178814

 View more versions of this product

Catalog No. AAJ6178814


Explore available promotions

Description

Description

Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

Solubility
Soluble in water (50 mg/ml at 25°C), ethanol (20 mg/ml), methanol, DMSO (50 mg/ml at 25°C), and phosphate buffered saline (50 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

DL-Isoproterenol hydrochloride
165°C to 175°C (decomposition)
C11H18ClNO3
25 g
14,5217
Soluble in water (50mg/ml at 25°C),ethanol (20mg/ml),methanol,DMSO (50mg/ml at 25°C),and phosphate buffered saline (50 mM).
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
247.72
247.72
Crystalline Powder
51-30-9
White to Yellow
MFCD00012603,MFCD00064548
3917363
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
IROWCYIEJAOFOW-UHFFFAOYNA-N
hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
5807
98%

Chemical Identifiers

51-30-9
247.72
IROWCYIEJAOFOW-UHFFFAOYNA-N
5807
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1
C11H18ClNO3
MFCD00012603,MFCD00064548
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso
hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 200-089-8

RTECSNumber : DO1925000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only