Learn More
DL-Isoproterenol hydrochloride, 98%, Thermo Scientific Chemicals
Standard selective beta-adrenoceptor agonist
$114.88 - $474.81
Chemical Identifiers
CAS | 51-30-9 |
---|---|
Molecular Formula | C11H18ClNO3 |
Molecular Weight (g/mol) | 247.72 |
MDL Number | MFCD00012603,MFCD00064548 |
InChI Key | IROWCYIEJAOFOW-UHFFFAOYNA-N |
Synonym | isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso |
PubChem CID | 5807 |
IUPAC Name | hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride |
SMILES | [H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6178806
|
Thermo Scientific Chemicals
J6178806 |
5 g |
Each for $114.88
|
|
|||||
AAJ6178814
|
Thermo Scientific Chemicals
J6178814 |
25 g |
Each for $474.81
|
|
|||||
Description
Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.
Solubility
Soluble in water (50 mg/ml at 25°C), ethanol (20 mg/ml), methanol, DMSO (50 mg/ml at 25°C), and phosphate buffered saline (50 mM).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Chemical Identifiers
51-30-9 | |
247.72 | |
IROWCYIEJAOFOW-UHFFFAOYNA-N | |
5807 | |
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 |
C11H18ClNO3 | |
MFCD00012603,MFCD00064548 | |
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso | |
hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride |
Specifications
51-30-9 | |
White to Yellow | |
MFCD00012603,MFCD00064548 | |
3917363 | |
isoprenaline hydrochloride, isoproterenol hydrochloride, isuprel, euspiran, isoprenaline hcl, isoproterenol hcl, aerolone, aerotrol, izadrin, vapo-iso | |
IROWCYIEJAOFOW-UHFFFAOYNA-N | |
hydrogen 4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol chloride | |
5807 | |
98% | |
DL-Isoproterenol hydrochloride |
165°C to 175°C (decomposition) | |
C11H18ClNO3 | |
5 g | |
14,5217 | |
Soluble in water (50mg/ml at 25°C),ethanol (20mg/ml),methanol,DMSO (50mg/ml at 25°C),and phosphate buffered saline (50 mM). | |
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(O)C(O)=C1 | |
247.72 | |
247.72 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-089-8
RTECSNumber : DO1925000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only