Learn More
DL-Menthol, 98+%
CAS: 89-78-1 | C10H20O | 156.269 g/mol
$61.96 - $455.45
Chemical Identifiers
| CAS | 89-78-1 |
|---|---|
| Molecular Formula | C10H20O |
| Molecular Weight (g/mol) | 156.269 |
| MDL Number | MFCD00001484 |
| InChI Key | NOOLISFMXDJSKH-IVZWLZJFSA-N |
| Synonym | menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol |
| PubChem CID | 6566020 |
| IUPAC Name | (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
| SMILES | CC1CCC(C(C1)O)C(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
AAA1809818
|
Thermo Scientific Chemicals
A1809818 |
50 g |
Each for $61.96
|
|
|||||
|
AAA1809822
|
Thermo Scientific Chemicals
A1809822 |
100 g |
Each for $88.10
|
|
|||||
|
AAA1809830
|
Thermo Scientific Chemicals
A1809830 |
250 g |
Each for $142.27
|
|
|||||
|
AAA180980B
|
Thermo Scientific Chemicals
A180980B |
1000 g |
Each for $455.45
|
|
|||||
Description
Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its cooling action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat bad breath. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. Spud and Kool brands) for both its flavor and also to reduce irritation, especially to the throat.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in alcohol, paraffin oil and water, 434.5 mg/L @ 25°C (est).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
| 89-78-1 | |
| 156.269 | |
| NOOLISFMXDJSKH-IVZWLZJFSA-N | |
| 6566020 | |
| CC1CCC(C(C1)O)C(C)C |
| C10H20O | |
| MFCD00001484 | |
| menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
| (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
Specifications
| 89-78-1 | |
| 0.89 | |
| 93°C (199°F) | |
| C10H20O | |
| MFCD00001484 | |
| 3194263 | |
| menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
| NOOLISFMXDJSKH-IVZWLZJFSA-N | |
| (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
| 6566020 | |
| ≥98% |
| 33°C to 36°C | |
| 212°C to 216°C | |
| Sweet | |
| 1.4615 | |
| 50 g | |
| 14,5837 | |
| Soluble in alcohol,paraffin oil and water,434.5mg/L @ 25°C (est). | |
| CC1CCC(C(C1)O)C(C)C | |
| 156.269 | |
| 156.27 | |
| DL-Menthol |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-939-0
RTECSNumber : OT0350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only