Learn More
DL-Menthol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 125395000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-Menthol | |
89-78-1 | |
Colorless to White | |
216.0°C | |
Authentic | |
Glass bottle | |
MFCD00001484 | |
0.89 | |
−2.00 (20.00°C c=10,EtOH) | |
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
NOOLISFMXDJSKH-IVZWLZJFSA-N | |
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
6566020 | |
99% |
99% | |
32.0°C to 36.0°C | |
0.8900g/mL | |
92°C | |
98.5% min. (GC) | |
C10H20O | |
500 mL | |
15, 5905 | |
− 2.00 | |
Solubility in water: insoluble. Other solubilities: freely soluble in glacial acetic acid,soluble in alcohol,chloroform and ether | |
CC1CCC(C(C1)O)C(C)C | |
156.27 | |
156.27 | |
Crystalline Solid |
Chemical Identifiers
89-78-1 | |
156.27 | |
NOOLISFMXDJSKH-IVZWLZJFSA-N | |
6566020 | |
CC1CCC(C(C1)O)C(C)C |
C10H20O | |
MFCD00001484 | |
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 201-939-
RTECSNumber : GW0554000
TSCA : TSCA
RUO – Research Use Only