missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Menthol, 99%, Thermo Scientific Chemicals

$60.33 - $360.73

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.27
MDL Number MFCD00001484
InChI Key NOOLISFMXDJSKH-IVZWLZJFSA-N
Synonym menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
PubChem CID 6566020
IUPAC Name (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC125390050
SDS View Documents
Thermo Scientific Chemicals
125390050
5 mL Glass bottle
Each for $60.33
 
AC125391000
SDS View Documents
Thermo Scientific Chemicals
125391000
100 mL Glass bottle
Each for $105.79
 
AC125395000
SDS View Documents
Thermo Scientific Chemicals
125395000
500 mL Glass bottle
Each for $360.73
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

89-78-1
156.27
NOOLISFMXDJSKH-IVZWLZJFSA-N
6566020
CC1CCC(C(C1)O)C(C)C
C10H20O
MFCD00001484
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol

Specifications

89-78-1
Colorless to White
216.0°C
Authentic
Glass bottle
MFCD00001484
0.89
−2.00 (20.00°C c=10,EtOH)
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
NOOLISFMXDJSKH-IVZWLZJFSA-N
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
6566020
99%
DL-Menthol, 99%
32.0°C to 36.0°C
0.8900g/mL
92°C
98.5% min. (GC)
C10H20O
5 mL
15, 5905
− 2.00
Solubility in water: insoluble. Other solubilities: freely soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
CC1CCC(C(C1)O)C(C)C
156.27
156.27
Crystalline Solid
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 201-939-

RTECSNumber : GW0554000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only