missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Menthol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 125390050

 View more versions of this product

Catalog No. AC125390050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

DL-Menthol
89-78-1
Colorless to White
216.0°C
Authentic
Glass bottle
MFCD00001484
0.89
−2.00 (20.00°C c=10,EtOH)
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
NOOLISFMXDJSKH-IVZWLZJFSA-N
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
6566020
99%
99%
32.0°C to 36.0°C
0.8900g/mL
92°C
98.5% min. (GC)
C10H20O
5 mL
15, 5905
− 2.00
Solubility in water: insoluble. Other solubilities: freely soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
CC1CCC(C(C1)O)C(C)C
156.27
156.27
Crystalline Solid

Chemical Identifiers

89-78-1
156.27
NOOLISFMXDJSKH-IVZWLZJFSA-N
6566020
CC1CCC(C(C1)O)C(C)C
C10H20O
MFCD00001484
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 201-939-

RTECSNumber : GW0554000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only