missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 137875000

 View more versions of this product

Catalog No. AC137875000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

DL-Tartaric acid
133-37-9
99.50%
HO2CCH(OH)CH(OH)CO2H
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
FEWJPZIEWOKRBE-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid
875
150.09
500 g
Authentic
Plastic bottle
200.0°C to 206.0°C
White
C4H6O6
MFCD00071626
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether, insoluble in chloroform
OC(C(O)C(O)=O)C(O)=O
150.09
CHEBI:15674
99.5%
210°C
0.5% max.
Crystalline Powder

Chemical Identifiers

133-37-9
150.09
FEWJPZIEWOKRBE-UHFFFAOYNA-N
875
2,3-dihydroxybutanedioic acid
C4H6O6
MFCD00071626
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
CHEBI:15674
OC(C(O)C(O)=O)C(O)=O
Videos
Safety and Handling

Safety and Handling

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only