missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Tartaric acid, 99.5%, Thermo Scientific Chemicals

$64.02 - $783.69

Chemical Identifiers

CAS 133-37-9
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00071626
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
PubChem CID 875
ChEBI CHEBI:15674
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC137871000
SDS View Documents
Thermo Scientific Chemicals
137871000
100 g Plastic bottle
Each for $64.02
 
AC137875000
SDS View Documents
Thermo Scientific Chemicals
137875000
500 g Plastic bottle
Each for $188.58
 
AC137870025
SDS View Documents
Thermo Scientific Chemicals
137870025
2.5 kg Plastic bottle
Each for $783.69
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

133-37-9
150.09
FEWJPZIEWOKRBE-UHFFFAOYNA-N
875
2,3-dihydroxybutanedioic acid
C4H6O6
MFCD00071626
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
CHEBI:15674
OC(C(O)C(O)=O)C(O)=O

Specifications

200.0°C to 206.0°C
White
C4H6O6
MFCD00071626
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether, insoluble in chloroform
OC(C(O)C(O)=O)C(O)=O
150.09
CHEBI:15674
99.5%
210°C
0.5% max.
Crystalline Powder
133-37-9
99.50%
HO2CCH(OH)CH(OH)CO2H
dl-tartaric acid, 2,3-dihydroxysuccinic acid, tartaric acid, racemic acid, uvic acid, traubensaure, paratartaric acid, paratartaric aicd, dl-tartrate, racemic tartaric acid
FEWJPZIEWOKRBE-UHFFFAOYNA-N
2,3-dihydroxybutanedioic acid
875
150.09
100 g
Authentic
Plastic bottle
DL-Tartaric acid
Videos
Safety and Handling

Safety and Handling

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only