missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Tropic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140550050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-Tropic acid | |
117.0°C to 120.0°C | |
Authentic | |
Glass bottle | |
C6H5CH(CH2OH)CO2H | |
5 g | |
15, 9958 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,practically insoluble in petroleum ether | |
OCC(C(O)=O)C1=CC=CC=C1 | |
166.18 | |
CHEBI:30765 | |
97% |
552-63-6 | |
White | |
96% min. (HPLC) | |
C9H10O3 | |
MFCD00004255 | |
10, 261 | |
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
JACRWUWPXAESPB-UHFFFAOYNA-N | |
3-hydroxy-2-phenylpropanoic acid | |
10726 | |
166.18 | |
Crystalline Powder |
Chemical Identifiers
552-63-6 | |
166.18 | |
JACRWUWPXAESPB-UHFFFAOYNA-N | |
10726 | |
3-hydroxy-2-phenylpropanoic acid |
C9H10O3 | |
MFCD00004255 | |
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
CHEBI:30765 | |
OCC(C(O)=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
EINECSNumber : 209-020-6
RUO – Research Use Only