missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Tropic acid, 97%, Thermo Scientific Chemicals
$35.97 - $35.97
Chemical Identifiers
CAS | 552-63-6 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00004255 |
InChI Key | JACRWUWPXAESPB-UHFFFAOYNA-N |
Synonym | tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid |
PubChem CID | 10726 |
ChEBI | CHEBI:30765 |
IUPAC Name | 3-hydroxy-2-phenylpropanoic acid |
SMILES | OCC(C(O)=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC140550050
|
Thermo Scientific Chemicals
140550050 |
5 g | Glass bottle |
Each for $35.97
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
552-63-6 | |
166.18 | |
JACRWUWPXAESPB-UHFFFAOYNA-N | |
10726 | |
3-hydroxy-2-phenylpropanoic acid |
C9H10O3 | |
MFCD00004255 | |
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
CHEBI:30765 | |
OCC(C(O)=O)C1=CC=CC=C1 |
Specifications
552-63-6 | |
White | |
96% min. (HPLC) | |
C9H10O3 | |
MFCD00004255 | |
10, 261 | |
tropic acid, dl-tropic acid, 2-phenylhydracrylic acid, 3-hydroxy-2-phenylpropionic acid, +--tropic acid, tropate, alpha-hydroxymethyl benzeneacetic acid, beta-hydroxyhydratropic acid, hydracrylic acid, 2-phenyl, alpha-hydroxymethyl phenylacetic acid | |
JACRWUWPXAESPB-UHFFFAOYNA-N | |
3-hydroxy-2-phenylpropanoic acid | |
10726 | |
166.18 | |
Crystalline Powder |
117.0°C to 120.0°C | |
Authentic | |
Glass bottle | |
C6H5CH(CH2OH)CO2H | |
5 g | |
15, 9958 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in benzene,practically insoluble in petroleum ether | |
OCC(C(O)=O)C1=CC=CC=C1 | |
166.18 | |
CHEBI:30765 | |
97% | |
DL-Tropic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
EINECSNumber : 209-020-6
RUO – Research Use Only