Learn More
Dodecane, mixture of isomers, Thermo Scientific Chemicals
Dodecane, C12H26, CAS Number-13475-82-6, heptane,2,2,4,6,6-pentamethyl, 2,2,4,6,6-pentamethyl-heptane, heptane, 2,2,4,6,6-pentamethyl, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, permethyl 99a, 1L, 170.0 deg.C to 195.0 deg.C, CHEBI:77509, Undesignated, 0.7510g/mL, 236-757-, 44 deg.C, 170.34
Supplier: Thermo Scientific Chemicals 365770010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dodecane | |
-60.0°C | |
170.0°C to 195.0°C | |
Authentic | |
C12H26 | |
MFCD00008969 | |
0.751 | |
Solubility in water: not miscible. | |
CC(CC(C)(C)C)CC(C)(C)C | |
170.34 | |
CHEBI:77509 | |
Liquid |
13475-82-6 | |
0.7510g/mL | |
44°C | |
Glass bottle | |
1.4190 to 1.4230 | |
1 L | |
heptane, 2,2,4,6,6-pentamethyl, heptane,2,2,4,6,6-pentamethyl, permethyl 99a, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, 2,2,4,6,6-pentamethyl-heptane | |
VKPSKYDESGTTFR-UHFFFAOYSA-N | |
2,2,4,6,6-pentamethylheptane | |
26058 | |
170.34 |
Chemical Identifiers
13475-82-6 | |
170.34 | |
VKPSKYDESGTTFR-UHFFFAOYSA-N | |
CHEBI:77509 | |
CC(CC(C)(C)C)CC(C)(C)C |
C12H26 | |
MFCD00008969 | |
26058 | |
2,2,4,6,6-pentamethylheptane |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
EINECSNumber : 236-757-
RUO – Research Use Only