missing translation for 'onlineSavingsMsg'
Learn More

Dodecane, mixture of isomers

Dodecane, C12H26, CAS Number-13475-82-6, heptane,2,2,4,6,6-pentamethyl, 2,2,4,6,6-pentamethyl-heptane, heptane, 2,2,4,6,6-pentamethyl, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, permethyl 99a, 1L, 170.0 deg.C to 195.0 deg.C, CHEBI:77509, Undesignated, 0.7510g/mL, 236-757-, 44 deg.C, 170.34 | CAS: 13475-82-6 | C12H26 | 170.34 g/mol

$107.99 - $1272.70

Chemical Identifiers

CAS 13475-82-6
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
MDL Number MFCD00008969
InChI Key VKPSKYDESGTTFR-UHFFFAOYSA-N
PubChem CID 26058
ChEBI CHEBI:77509
IUPAC Name 2,2,4,6,6-pentamethylheptane
SMILES CC(CC(C)(C)C)CC(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC365770010
SDS View Documents
Thermo Scientific Chemicals
365770010
1 L Glass bottle
Each for $107.99
Only null left
 
AC365770050
SDS View Documents
Thermo Scientific Chemicals
365770050
5 L Plastic drum
Each for $342.76
Only null left
 
AC365770250
SDS View Documents
Thermo Scientific Chemicals
365770250
25 L Metal drum
Each for $1,272.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13475-82-6
170.34
VKPSKYDESGTTFR-UHFFFAOYSA-N
CHEBI:77509
CC(CC(C)(C)C)CC(C)(C)C
C12H26
MFCD00008969
26058
2,2,4,6,6-pentamethylheptane

Specifications

13475-82-6
0.7510g/mL
44°C
Glass bottle
1.4190 to 1.4230
1 L
heptane, 2,2,4,6,6-pentamethyl, heptane,2,2,4,6,6-pentamethyl, permethyl 99a, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, 2,2,4,6,6-pentamethyl-heptane
VKPSKYDESGTTFR-UHFFFAOYSA-N
2,2,4,6,6-pentamethylheptane
26058
170.34
Dodecane
-60.0°C
170.0°C to 195.0°C
Authentic
C12H26
MFCD00008969
0.751
Solubility in water: not miscible.
CC(CC(C)(C)C)CC(C)(C)C
170.34
CHEBI:77509
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 236-757-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only