missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dodecane, mixture of isomers, Thermo Scientific Chemicals

Dodecane, C12H26, CAS Number-13475-82-6, heptane,2,2,4,6,6-pentamethyl, 2,2,4,6,6-pentamethyl-heptane, heptane, 2,2,4,6,6-pentamethyl, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, permethyl 99a, 1L, 170.0 deg.C to 195.0 deg.C, CHEBI:77509, Undesignated, 0.7510g/mL, 236-757-, 44 deg.C, 170.34

Supplier:  Thermo Scientific Chemicals 365770250

 View more versions of this product

Catalog No. AC365770250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dodecane
-60.0°C
170.0°C to 195.0°C
Authentic
C12H26
MFCD00008969
0.751
Solubility in water: not miscible.
CC(CC(C)(C)C)CC(C)(C)C
170.34
CHEBI:77509
Liquid
13475-82-6
0.7510g/mL
44°C
Metal drum
1.4190 to 1.4230
25 L
heptane, 2,2,4,6,6-pentamethyl, heptane,2,2,4,6,6-pentamethyl, permethyl 99a, acmc-1c04u, vkpskydesgttfr-uhfffaoysa, 2,2,4,6,6-pentamethyl-heptane
VKPSKYDESGTTFR-UHFFFAOYSA-N
2,2,4,6,6-pentamethylheptane
26058
170.34

Chemical Identifiers

13475-82-6
170.34
VKPSKYDESGTTFR-UHFFFAOYSA-N
CHEBI:77509
CC(CC(C)(C)C)CC(C)(C)C
C12H26
MFCD00008969
26058
2,2,4,6,6-pentamethylheptane
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o

GHS Signal Word: Danger

EINECSNumber : 236-757-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only