missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dodecylbenzene sulfonic acid, mixture of C10-C13 isomers, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 325905000

 View more versions of this product

Catalog No. AC325905000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dodecylbenzene sulfonic acid
85536-14-7
Brown
315.0°C
Authentic
CH3(CH2)xC6H4SO3Hx=9to12
MFCD00066677
1.06
Solubility in water: 10g/L (20°C). Other solubilities: soluble in alkaline solutions, soluble in ethanol
≥96% (sulfonic acid, M.W. 322)
Mixture of C10-C13 Isomers
-10.0°C
1.0600g/mL
210°C
Glass bottle
1.5035 to 1.5065
500 g
dodecylbenzenesulphonic acid, benzenesulfonic acid, 2-dodecyl, dodecyl benzene sulfonic acid, o-dodecylbenzenesulfonic acid, unii-qmo4a07h35, dodecyl benzenesulfonic acid, dodecylbenzene sulfonic acid, dodecylbenzenesulfonicacid, 2-dodecylbenzenesulphonic acid, dodecyl benzene sulphonic acid
2400 mPa.s
Viscous Liquid

Chemical Identifiers

85536-14-7
MFCD00066677
dodecylbenzenesulphonic acid, benzenesulfonic acid, 2-dodecyl, dodecyl benzene sulfonic acid, o-dodecylbenzenesulfonic acid, unii-qmo4a07h35, dodecyl benzenesulfonic acid, dodecylbenzene sulfonic acid, dodecylbenzenesulfonicacid, 2-dodecylbenzenesulphonic acid, dodecyl benzene sulphonic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 287-494-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only