Learn More
Dodecylbenzene sulfonic acid, mixture of C10-C13 isomers, Thermo Scientific Chemicals
$315.69 - $565.37
Chemical Identifiers
CAS | 85536-14-7 |
---|---|
MDL Number | MFCD00066677 |
Synonym | dodecylbenzenesulphonic acid, benzenesulfonic acid, 2-dodecyl, dodecyl benzene sulfonic acid, o-dodecylbenzenesulfonic acid, unii-qmo4a07h35, dodecyl benzenesulfonic acid, dodecylbenzene sulfonic acid, dodecylbenzenesulfonicacid, 2-dodecylbenzenesulphonic acid, dodecyl benzene sulphonic acid |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC325905000
|
Thermo Scientific Chemicals
325905000 |
500 g | Glass bottle |
N/A
|
|
|||||
AC325900010
|
Thermo Scientific Chemicals
325900010 |
1 kg | Glass bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
85536-14-7 | |
MFCD00066677 | |
dodecylbenzenesulphonic acid, benzenesulfonic acid, 2-dodecyl, dodecyl benzene sulfonic acid, o-dodecylbenzenesulfonic acid, unii-qmo4a07h35, dodecyl benzenesulfonic acid, dodecylbenzene sulfonic acid, dodecylbenzenesulfonicacid, 2-dodecylbenzenesulphonic acid, dodecyl benzene sulphonic acid |
Specifications
85536-14-7 | |
Brown | |
315.0°C | |
Authentic | |
CH3(CH2)xC6H4SO3Hx=9to12 | |
MFCD00066677 | |
1.06 | |
Solubility in water: 10g/L (20°C). Other solubilities: soluble in alkaline solutions, soluble in ethanol | |
≥96% (sulfonic acid, M.W. 322) | |
Dodecylbenzene sulfonic acid, Mixture of C10-C13 Isomers |
-10.0°C | |
1.0600g/mL | |
210°C | |
Glass bottle | |
1.5035 to 1.5065 | |
500 g | |
dodecylbenzenesulphonic acid, benzenesulfonic acid, 2-dodecyl, dodecyl benzene sulfonic acid, o-dodecylbenzenesulfonic acid, unii-qmo4a07h35, dodecyl benzenesulfonic acid, dodecylbenzene sulfonic acid, dodecylbenzenesulfonicacid, 2-dodecylbenzenesulphonic acid, dodecyl benzene sulphonic acid | |
2400 mPa.s | |
Viscous Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 287-494-3
RUO – Research Use Only