Thermo Scientific™ Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
$89.24 - $248.39
Specifications
Format Strips of 8 Flat Caps
View More Specs
Products 3
Catalog Number Mfr. No. Color Format Quantity Price Quantity  
Catalog Number Mfr. No. Color Format Quantity Price Quantity  
AB-0783
View Documents
Thermo Scientific™
AB0783
Clear Strips of 8 Flat Caps 120/Cs.
Case of 120 Each for $89.24
AB-0784
View Documents
Thermo Scientific™
AB0784
Clear Strips of 8 Flat Caps 250/Cs.
Case of 120 Each for $132.00
AB-0786
View Documents
Thermo Scientific™
AB0786
Natural Strips of 8 Flat Caps 25 packs of 12
Case of 300 Each for $248.39
Description
Description
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Specifications
Specifications
Strips of 8 Flat Caps
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'