Thermo Scientific™ Domed PCR Caps, strips of 8
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Supplier: Thermo Scientific™ AB0386
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.Highlights:
- Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
- Large end tabs for strip removal (12 strips only)
Certified free from human DNA, RNases, and DNases.
Specifications
Clear | |
8 | |
Strips of 8 Domed Caps | |
Yes |
Polypropylene | |
10 packs of 12 | |
PCR Plate Cap Strip | |
96-well Plate |