Thermo Scientific™ Domed PCR Caps, strips of 8
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
$87.26 - $195.79
Specifications
Color Clear
Material Polypropylene
No. of Wells 8
Format Strips of 8 Domed Caps
Product Type PCR Plate Cap Strip
View More Specs
Products 2
Catalog Number Mfr. No. Format Quantity Price Quantity  
Catalog Number Mfr. No. Format Quantity Price Quantity  
AB-0265
View Documents
Thermo Scientific™
AB0265
Strips of 8 Domed Caps 250 strips N/A
AB-0386
View Documents
Thermo Scientific™
AB0386
Strips of 8 Domed Caps 10 packs of 12 N/A
Description
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
Clear
8
PCR Plate Cap Strip
96-well Plate
Polypropylene
Strips of 8 Domed Caps
Yes
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'