Learn More
Domiphen bromide, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1293014
Description
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDomiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.
Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Domiphen bromide | |
117°C to 118°C | |
MFCD00011769 | |
3922742 | |
14,3415 | |
Soluble in ethanol,acetone,ethyl acetate,chloroform and water. Slightly soluble in benzene. | |
CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-] | |
414.472 | |
414.48 |
538-71-6 | |
C22H40BrNO | |
25 g | |
Hygroscopic | |
domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r | |
OJIYIVCMRYCWSE-UHFFFAOYSA-M | |
dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide | |
10866 | |
97% |
Chemical Identifiers
538-71-6 | |
414.472 | |
OJIYIVCMRYCWSE-UHFFFAOYSA-M | |
10866 | |
CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-] |
C22H40BrNO | |
MFCD00011769 | |
domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r | |
dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 208-702-0
RTECSNumber : BQ2030000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only