missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Domiphen bromide, 97%, Thermo Scientific Chemicals

$94.96 - $261.14

Chemical Identifiers

CAS 538-71-6
Molecular Formula C22H40BrNO
Molecular Weight (g/mol) 414.472
MDL Number MFCD00011769
InChI Key OJIYIVCMRYCWSE-UHFFFAOYSA-M
Synonym domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
PubChem CID 10866
IUPAC Name dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
SMILES CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1293014
View Documents
Thermo Scientific Chemicals
L1293014
25 g
Each for $94.96
 
AAL1293022
View Documents
Thermo Scientific Chemicals
L1293022
100 g
Each for $261.14
 
Description

Description

Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domiphen bromide is employed as a phase transfer catalyst in synthetic chemistry. It is used as a quaternary ammonium cation with cationic surfactant properties. Further, it is used as an antiseptic and an active pharmaceutical ingredient.

Solubility
Soluble in ethanol, acetone, ethyl acetate, chloroform and water. Slightly soluble in benzene.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

538-71-6
414.472
OJIYIVCMRYCWSE-UHFFFAOYSA-M
10866
CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]
C22H40BrNO
MFCD00011769
domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide

Specifications

538-71-6
C22H40BrNO
25 g
Hygroscopic
domiphen bromide, bradasol, bradonit, bradoral, fungitex, oradol, phenododecinium bromide, bradosol bromide, fungitex r
OJIYIVCMRYCWSE-UHFFFAOYSA-M
dodecyl-dimethyl-(2-phenoxyethyl)azanium;bromide
10866
97%
117°C to 118°C
MFCD00011769
3922742
14,3415
Soluble in ethanol,acetone,ethyl acetate,chloroform and water. Slightly soluble in benzene.
CCCCCCCCCCCC[N+](C)(C)CCOC1=CC=CC=C1.[Br-]
414.472
414.48
Domiphen bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 208-702-0

RTECSNumber : BQ2030000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only