missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Accuform Signs DOT Shipping Labels
to place proper identification and classification of hazardous material during transporting and shipping
Supplier: Accuform Signs MSL801PS2
Specifications
Self-adhesive | |
Paper | |
Corrosive 8 | |
Graphic | |
4 in. | |
10.16 cm |
Label | |
DOT Shipping Label | |
The durable adhesive will stick to most flat, clean surfaces, Adhesive Vinyl signs will work for short term outdoor applications and in moderate temperature | |
250/Pk. | |
4 in. |