missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Accuform Signs DOT Shipping Labels
to place proper identification and classification of hazardous material during transporting and shipping
$43.08 - $70.68
Specifications
Adhesive Type | Self-adhesive |
---|---|
Format | Label |
Product Type | DOT Shipping Label |
Quantity | 250/Pk. |
Height (English) | 4 in. |
Catalog Number | Mfr. No. | Quantity | Display Text | Material | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Display Text | Material | Price | Quantity | |||
19-804-962
|
Accuform Signs
MSL301EV2 |
250/Pk. | Flammable Liquid 3 | Vinyl |
Each for $70.68
|
|
|||
19-809-835
|
Accuform Signs
MSL202PS2 |
250/Pk. | Flammable Gas 2 | Paper |
Pack of 250 Each for $44.97
|
|
|||
19-809-837
|
Accuform Signs
MSL801PS2 |
250/Pk. | Corrosive 8 | Paper |
Pack of 250 Each for $43.08
|
|
|||
19-809-836
|
Accuform Signs
MSL301PS2 |
250/Pk. | Flammable Liquid 3 | Paper |
Pack of 250 Each for $44.97
|
|
|||
Specifications
Self-adhesive | |
DOT Shipping Label | |
4 in. |
Label | |
250/Pk. | |
10.16 cm |