missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ DOT Vehicle Placards: (Misc. Hazardous Materials) 9
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
Supplier: Brady™ 63421
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
Acrylic, Permanent, Cold-temperature | |
Pressure-sensitive Vinyl (B-946) | |
Misc. Hazardous Materials, 9 | |
1 Ea. | |
10.75 x 10.75 in. |
Black, White | |
Vehicle Placard | |
Material Handling, Shipping & Receiving Signs & Labels | |
Diamond | |
0.004 in. |