missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ DOT Vehicle Placards: (Misc. Hazardous Materials) 9
Comply with DOT regulations by marking delivery vehicles carrying dangerous or hazardous materials. Brady™ DOT Vehicle Placards are available in a variety of materials.
$4.89 - $40.19
Specifications
Color | Black, White |
---|---|
Product Type | Vehicle Placard |
Display Text | Misc. Hazardous Materials, 9 |
For Use With (Application) | Material Handling, Shipping & Receiving Signs & Labels |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
19-111-332
|
Brady™
63421 |
Pressure-sensitive Vinyl (B-946) |
Each for $16.96
|
|
|||||
19-111-340
|
Brady™
63434 |
Fiberglass (B-120) |
Each for $40.19
|
|
|||||
19-111-356
|
Brady™
63455 |
Polycoated Tagstock (B-101) |
Each for $4.89
|
|
|||||
Description
- Available in three different materials Pressure Sensitive Vinyl, Polycoated Tagstock and Premium Fiberglass
Specifications
Black, White | |
Misc. Hazardous Materials, 9 | |
1 Ea. | |
10.75 x 10.75 in. |
Vehicle Placard | |
Material Handling, Shipping & Receiving Signs & Labels | |
Diamond |