missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LaMotte™ DPD Free Chlorine Reagent Refills

The liquid alternative to DPD tablet refills

Supplier:  LaMotte™ P6743CNG

 View more versions of this product

Catalog No. 22-278-647


Explore available promotions

Alert:

Keep out of reach of children. May irritate eyes and skin. Harmful if swallowed.


Description

Description

  • DPD £1A and DPD £1B test Free Available Chlorine (Cl)
  • DPD £3 is added to the reacted sample to measure Total Cl
  • Comes in 30mL and 60mL sizes

DPD = N, N-Diethyl-1, 4-phenylene-diamine sulfate

Use with existing LaMotte chlorine comparators or colorimeters. For each test, all three reagents are added to the same sample. Ideal for testing drinking water, food and beverage, pool and spa, industrial water boiling and cooling, laundry and sanitation, water and wastewater.

Specifications

Specifications

30 mL
LaMotte™ colorimeters and comparators
DPD 3 Liquid Reagent
Measurement of total chlorine, provides quicker results and clearer colors than tablets
Each
1 oz.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions